missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-tert-butyl-2-methoxybenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-7052592
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1516-95-6
C15H24O
220.36
MFCD20483412
OGIJRUWUWSZKAV-UHFFFAOYSA-N
1,3-di-tert-butyl-2-methoxybenzene, benzene, 1,3-bis 1,1-dimethylethyl-2-methoxy, 2,6-di-tert-butylanisole
12794694
1,3-ditert-butyl-2-methoxybenzene
COC1=C(C=CC=C1C(C)(C)C)C(C)(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1516-95-6
C15H24O
220.36
MFCD20483412
OGIJRUWUWSZKAV-UHFFFAOYSA-N
1,3-di-tert-butyl-2-methoxybenzene, benzene, 1,3-bis 1,1-dimethylethyl-2-methoxy, 2,6-di-tert-butylanisole
12794694
1,3-ditert-butyl-2-methoxybenzene
COC1=C(C=CC=C1C(C)(C)C)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Boiling Point 124°C (9 mmHg)
Quantity 10 g
Formula Weight 220.35
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,3-Di-tert-butyl-2-methoxybenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'