missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose, 98%

Catalog Number p-7043187
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
144490-03-9
C13H18O9
318.278
MFCD00674547
IHNHAHWGVLXCCI-UMSGYPCISA-N
1,2,3,5-tetra-o-acetyl-beta-l-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-l-ribofuranose, 2s,3s,4s,5r-3,4,5-tris acetyloxy oxolan-2-yl methyl acetate, beta-l-ribofuranose 1,2,3,5-tetra-o-acetate, beta-l-ribofuranose tetraacetate, 1,2,3,5-tetra-o-acetyl-?-l-ribofuranose, 2s,3s,4s,5r-3,4,5-triacetyloxyoxolan-2-yl methyl acetate, 2r,3s,4s,5s-5-acetoxymethyl tetrahydrofuran-2,3,4-triyl triacetate
69431757
[(2S,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
This item is not returnable. View return policy
144490-03-9
C13H18O9
318.278
MFCD00674547
IHNHAHWGVLXCCI-UMSGYPCISA-N
1,2,3,5-tetra-o-acetyl-beta-l-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-l-ribofuranose, 2s,3s,4s,5r-3,4,5-tris acetyloxy oxolan-2-yl methyl acetate, beta-l-ribofuranose 1,2,3,5-tetra-o-acetate, beta-l-ribofuranose tetraacetate, 1,2,3,5-tetra-o-acetyl-?-l-ribofuranose, 2s,3s,4s,5r-3,4,5-triacetyloxyoxolan-2-yl methyl acetate, 2r,3s,4s,5s-5-acetoxymethyl tetrahydrofuran-2,3,4-triyl triacetate
69431757
[(2S,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 80°C to 83°C
Color White
Quantity 1 g
Optical Rotation +12.7° (c=2.4 in Chloroform)
Formula Weight 318.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'