missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose, 98%

Catalog Number p-7043263
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4163-65-9
C16H22O11
390.341
MFCD00069798
LPTITAGPBXDDGR-OWYFMNJBSA-N
a-d-mannose pentaacetate, alpha-d-mannose pentaacetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-mannopyranose, alpha-d-mannopyranose pentaacetate, 1-o,2-o,3-o,4-o,6-o-pentaacetyl-a-d-mannopyranose, man oac 5, a-d-mannosepentaacetate, pubchem22277, 1,2,3,4,6-penta-o-acetyl-, a-d-mannopyranose, 1,2,3,4,6-penta-o-acetyl-a-d-mannopyranose
11741089
[(2R,3R,4S,5S,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
This item is not returnable. View return policy
4163-65-9
C16H22O11
390.341
MFCD00069798
LPTITAGPBXDDGR-OWYFMNJBSA-N
a-d-mannose pentaacetate, alpha-d-mannose pentaacetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-mannopyranose, alpha-d-mannopyranose pentaacetate, 1-o,2-o,3-o,4-o,6-o-pentaacetyl-a-d-mannopyranose, man oac 5, a-d-mannosepentaacetate, pubchem22277, 1,2,3,4,6-penta-o-acetyl-, a-d-mannopyranose, 1,2,3,4,6-penta-o-acetyl-a-d-mannopyranose
11741089
[(2R,3R,4S,5S,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

α-D-Mannose pentaacetate is used for glycosylation in a study that assessed novel synthetic inhibitors of selectin-mediated cell adhesion. It has also been used in a study to investigate stereospecific entry to spiroketal glycosides using alkylidenecarbene C-H insertion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-D-Mannose pentaacetate is used for glycosylation in a study that assessed novel synthetic inhibitors of selectin-mediated cell adhesion. It has also been used in a study to investigate stereospecific entry to spiroketal glycosides using alkylidenecarbene C-H insertion.

Solubility
Soluble in chloroform

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
Solubility Information Soluble in chloroform
Optical Rotation +54° (c=1 in Chloroform)
Formula Weight 390.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'