missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-2-phenyl-3-butyn-2-ol, 96%, Thermo Scientific Chemicals

Catalog Number p-7033902
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
99727-20-5
C10H7F3O
200.16
MFCD00792447
WWIJKFJKIOQDKI-UHFFFAOYSA-N
1,1,1-trifluoro-2-phenyl-3-butyn-2-ol, 2-phenyl-1,1,1-trifluorobut-3-yn-2-ol, 2-phenyl-1.1.1-trifluorobut-3-yn-2-ol, 1,1,1-trifluoro-2-phenyl-3-butyne-2-ol, 1,1,1-trifluoro-2-phenyl-but-3-yn-2-ol, benzenemethanol,a-ethynyl-a-trifluoromethyl, 2s-1,1,1-trifluoro-2-phenyl-but-3-yn-2-ol, 1,1,1-tris fluoranyl-2-phenyl-but-3-yn-2-ol
2734575
1,1,1-trifluoro-2-phenylbut-3-yn-2-ol
C#CC(C1=CC=CC=C1)(C(F)(F)F)O
This item is not returnable. View return policy
99727-20-5
C10H7F3O
200.16
MFCD00792447
WWIJKFJKIOQDKI-UHFFFAOYSA-N
1,1,1-trifluoro-2-phenyl-3-butyn-2-ol, 2-phenyl-1,1,1-trifluorobut-3-yn-2-ol, 2-phenyl-1.1.1-trifluorobut-3-yn-2-ol, 1,1,1-trifluoro-2-phenyl-3-butyne-2-ol, 1,1,1-trifluoro-2-phenyl-but-3-yn-2-ol, benzenemethanol,a-ethynyl-a-trifluoromethyl, 2s-1,1,1-trifluoro-2-phenyl-but-3-yn-2-ol, 1,1,1-tris fluoranyl-2-phenyl-but-3-yn-2-ol
2734575
1,1,1-trifluoro-2-phenylbut-3-yn-2-ol
C#CC(C1=CC=CC=C1)(C(F)(F)F)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 200.16
Percent Purity 96%
Chemical Name or Material 1,1,1-Trifluoro-2-phenyl-3-butyn-2-ol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'