Bottletop Filter
Bottletop Filters
Bottletop filters are a practical way to streamline vacuum filtration of media, buffers, and aqueous solutions directly from standard bottles. Explore trusted options from Merck, Thermo Scientific Nalgene, Whatman, Corning, and Fisherbrand, including common pore sizes and membrane choices for routine sterilising filtration or clarification. Use brand filters to quickly compare formats and select the best fit for your workflow.
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.
    Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
    Fisherbrand™ Disposable PES Bottle Top Filters
    Increase sample protection with Fisherbrand Sterile Disposable Bottle Top Filters with PES Membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.
      Fisherbrand™ Disposable PES Bottle Top Filters
      Merck Stericup™ Quick Release-GP Sterile Vacuum Filtration Systems
      For sterile filter applications. Stericup Quick Release-GP Sterile Vacuum Filtration System.
        Merck Stericup™ Quick Release-GP Sterile Vacuum Filtration Systems
        Complete Your Order - Great Deals
        Vacuum Pumps
        Vaccum pumps create a partial vacuum for many applications including rotary evaporation, liquid transfer, distillation, filtration, freeze drying, degassing, and so on. In our portfolio you will also find pumps with a large variety of pressure ranges and different voltages, as well as vacuum systems.
        Vacuum Pumps
        Round Glass Bottles
        Glass bottles are inert and can provide better chemical compatibility than any kind of plastic, resulting in better sample integrity. Our collection of round glass bottles includes clear glass for content visibility, plastic/coated glass for extra resistance, and amber glass bottles for UV protection.
        Round Glass Bottles
        Syringeless Filters and Syringe Filters
        Syringeless filters and syringe filters are commonly constructed from PVDF or PTFE. They are used to filter impurities such as fine particles from small to medium sample volumes prior to use and are suitable for both aqueous and aqueous-organic samples.
        Syringeless Filters and Syringe Filters
        FAQ
        Bottletop filters are used for quick vacuum filtration of lab liquids such as media, buffers, and aqueous solutions directly from a bottle. They help streamline sterile filtration or clarification steps and reduce handling time. Common choices include Merck and Thermo Scientific Nalgene systems, plus Corning, Whatman, and Fisherbrand options.
        Choose pore size based on your goal: ~0.22 µm is commonly used for sterilising filtration of media and buffers, while ~0.45 µm is often used for clarification when sterility is not required. Compare equivalent formats across Merck, Corning, Fisherbrand, Whatman, and Thermo Scientific Nalgene to match your workflow.
        Membrane choice depends on chemistry and sample sensitivity. PES is widely used for aqueous solutions and offers good flow with low protein binding, while PTFE suits harsher solvents and nylon provides broad compatibility. Check the product description and compare trusted brands such as Merck, Corning, Whatman, Fisherbrand, and Thermo Scientific Nalgene.
        To reduce clogging, pre-clarify samples (settle or prefilter), avoid overloading with particulates, and choose the right membrane and format for viscosity. For challenging samples, consider a staged approach or larger surface area where available. Bottletop systems from Merck, Corning, and Thermo Scientific Nalgene are commonly used in routine workflows.
        Use the Brand Filters section to narrow results instantly by manufacturer. You can filter the Bottletop Filters category to view Merck, Thermo Scientific Nalgene, Whatman, Corning, or Fisherbrand ranges without leaving the page, making it easier to compare similar pore sizes, formats, and membranes side by side.
        Accessibility = 15 indicates products are set up across all supported countries, helping ensure consistent availability and merchandising across the EU. This page’s product tables only include Accessibility = 15 items, so selections are suitable for multi-country campaigns. You’ll still see a strong mix across key brands like Merck, Thermo Scientific Nalgene, Corning, Whatman, and Fisherbrand.
        Top Brands