accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62285-58-9

CH3H3CHO

CAS RN 62285-58-9

IUPAC Name: (2,6-dimethylphenyl)methanol
Synonyms: (2,6-dimethylphenyl)methanol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: JPEYJQDKTDVJSZ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1CO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results