accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62252-26-0

H3CONH2OOCl

CAS RN 62252-26-0

IUPAC Name: 7-amino-4-chloro-3-methoxy-1H-isochromen-1-one
Synonyms: 7-amino-4-chloro-3-methoxyisochromen-1-one
Molecular Weight (g/mol): 225.63
Molecular Formula: C10H8ClNO3
InChi Key: AMDGKLWVCUXONP-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C2=CC=C(N)C=C2C(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results