CAS RN 620113-73-7
CAS RN 620113-73-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CCG-63808, MedChemExpress
MedChemExpress CCG-63808 is a reversible inhibitor of regulator of G-protein signaling (RGS) proteins.