CAS RN 61849-14-7
CAS RN 61849-14-7
IUPAC Name:
sodium 5-[(2Z,3aR,4R,5R,6aS)-5-hydroxy-4-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-hexahydro-2H-cyclopenta[b]furan-2-ylidene]pentanoate
Synonyms:
sodium prostaglandin I2(1-)
Molecular Weight (g/mol):
374.45
Molecular Formula:
C20H31NaO5
InChi Key:
LMHIPJMTZHDKEW-XQYLJSSYSA-M
SMILES:
[Na+].CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)C[C@@H]2O\C(C[C@H]12)=C/CCCC([O-])=O