accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 61825-94-3

(R)(R)NNPtOOOO

CAS RN 61825-94-3

IUPAC Name: platinum(2+) cyclohexane-1,2-diamine oxalate
Synonyms: oxaliplatin
Molecular Weight (g/mol): 397.29
Molecular Formula: C8H14N2O4Pt
InChi Key: ZROHGHOFXNOHSO-UHFFFAOYNA-L
SMILES: [Pt++].[O-]C(=O)C([O-])=O.NC1CCCCC1N
Percent Purity 
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results