CAS RN 374790-92-8
CAS RN 374790-92-8
IUPAC Name:
(pentan-3-yl)(1-phenylethyl)amine hydrochloride
Synonyms:
pentan-3-yl(1-phenylethyl)amine hydrochloride
Molecular Weight (g/mol):
227.78
Molecular Formula:
C13H22ClN
InChi Key:
RKPRPVRHVBFULB-UHFFFAOYNA-N
SMILES:
Cl.CCC(CC)NC(C)C1=CC=CC=C1
1
–
1
of
1
results
1
(S)-(-)-N-(3-Pentyl)-1-phenylethylamine hydrochloride, 99%, Thermo Scientific™
CAS: 374790-92-8 Molecular Formula: C13H22ClN Molecular Weight (g/mol): 227.78 MDL Number: MFCD06199157 InChI Key: RKPRPVRHVBFULB-UHFFFAOYNA-N Synonym: s---n-3-pentyl-1-phenylethylamine hydrochloride,s-n-1-phenylethyl pentan-3-amine hydrochloride,pentan-3-yl 1s-1-phenylethyl amine hydrochloride PubChem CID: 22831494 IUPAC Name: N-[(1S)-1-phenylethyl]pentan-3-amine;hydrochloride SMILES: Cl.CCC(CC)NC(C)C1=CC=CC=C1