accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 150812-13-8

H3COOHNNHFNH2HClHCl

CAS RN 150812-13-8

IUPAC Name: ethyl N-(2-amino-4-{[(4-fluorophenyl)methyl]amino}phenyl)carbamate dihydrochloride
Synonyms: retigabine dihydrochloride
Molecular Weight (g/mol): 376.25
Molecular Formula: C16H20Cl2FN3O2
InChi Key: WSGFOWNASITQHJ-UHFFFAOYSA-N
SMILES: Cl.Cl.CCOC(=O)NC1=C(N)C=C(NCC2=CC=C(F)C=C2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results