accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1152-61-0

HOONHOOOHO

CAS RN 1152-61-0

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}butanedioic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}butanedioic acid
Molecular Weight (g/mol): 267.24
Molecular Formula: C12H13NO6
InChi Key: XYXYXSKSTZAEJW-UHFFFAOYNA-N
SMILES: OC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)