CAS RN 91575-26-7
CAS RN 91575-26-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
MedChemExpress GRGDSPC
MedChemExpress GRGDSPC, a 7-amino acid peptide, is a thiolated cell adhesion peptide.

2
MedChemExpress GRGDSPC
MedChemExpress GRGDSPC, a 7-amino acid peptide, is a thiolated cell adhesion peptide.

3
MedChemExpress GRGDSPC
MedChemExpress GRGDSPC, a 7-amino acid peptide, is a thiolated cell adhesion peptide.
