CAS RN 910808-12-7
CAS RN 910808-12-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Netupitant metabolite Monohydroxy Netupitant, MedChemExpress
MedChemExpress Monohydroxy Netupitant is the metabolite of Netupitant, which is a highly selective NK1 receptor antagonist.
