Learn More
Karl Hecht™ Watch Glasses
These watch glasses are useful for evaporating liquids, holding solids, and heating small amounts of a sample.
Brand:  Karl Hecht™ 40010014
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10339441
  • 3.50 € / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12 341
  • With fused rim
  • Manufactured from glass
  • Available in different diameters
Specifications
Specifications
Fused rim
Round
Holding, Evaporation, Heating
Glass
Watch Glass
14 cm