Learn More
SCHOTT AG Lighting and Imaging™ Twistable Polarization Filter for Focusing Lens
This Twistable Polarization Filter for Focusing Lens allows better viewing of polymeric structure properties in many materials. For use with focusing lens 158 215.
Brand: SCHOTT AG Lighting and Imaging™ 158206
Additional Details : Weight : 0.25000kg
Description
- Polarization filter.
- Twistable.
- Polarized light makes properties of the materials visible (polymeric structures).
- For use with focusing lens 158215.
Specifications
Polarization Filter | |
Stereo Microscopy Illumination | |
Focusing Lens 158215 |