Learn More
Gilson™ M-384 Well Adapter with Positioning by Wheel
An accessory allowing for the use of 384-well microplates with positioning done by wheel.
Brand:  Gilson™ F1077602
Additional Details : Weight : 0.60000kg
Product Code. 15893182
  • 2235.00 € / Each
In Stock
to see stock.
Add to basket
Description
Description
This adapter is an accessory for the PLATEMASTER™ that allows use of 384-well microplates with positioning done by wheel.
Specifications
Specifications
Pipetter Accessory
ISO 9001
Polypropylene
PLATEMASTER™
Not Autoclavable
White
Liquid Handling
1 Year