Learn More
Thermo Scientific™ Complete Manual Staining Assembly 100
Accommodate up to 19 slides with the Thermo Scientific™ Complete Staining Assembly 100, featuring a non-autoclavable glass dish.
22.45 € - 37.64 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12667696
|
Thermo Scientific™
101 |
Staining Dish Cover only for Assembly 100 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12657696
|
Thermo Scientific™
102 |
Staining Dish only for Assembly 100 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Accommodate up to 19 slides with the Thermo Scientific™ Complete Staining Assembly 100, featuring a non-autoclavable glass dish.
Includes:
- 109 rack/handle
- 102 dish
- 101 cover
Staining Rack 109:
Made from stainless steel and holds up to 19 slides. Maximum slide size is 3 x 2 in. (75 x 50mm).
Staining Dish 102:
Made from non-autoclavable glass with inside dimensions of 3.5 x 2.75 x 2.55 in. (90 x 70 x 65mm). Holds up to 250mL of solution with a fully loaded staining rack.
Made from stainless steel and holds up to 19 slides. Maximum slide size is 3 x 2 in. (75 x 50mm).
Staining Dish 102:
Made from non-autoclavable glass with inside dimensions of 3.5 x 2.75 x 2.55 in. (90 x 70 x 65mm). Holds up to 250mL of solution with a fully loaded staining rack.
Staining Dish Cover 101:
Made from stainless steel.