missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide, 98+%

Product Code. p-7023519
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423377 500 g 500g
11433377 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11423377 Supplier Thermo Scientific Alfa Aesar Supplier No. A17157.36
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 79-06-1 | C3H5NO | 71.08 g/mol

Acrylamide is used as a monomer in the synthesis of polyacrylamides, which finds application as water-soluble thickeners in waste water treatment, gel electrophoresis, ore processing and dyes production. In molecular biology, it is used as a carrier as well as involved in the precipitation of small amounts of DNA. It plays an important role as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide is used as a monomer in the synthesis of polyacrylamides, which finds application as water-soluble thickeners in waste water treatment, gel electrophoresis, ore processing and dyes production. In molecular biology, it is used as a carrier as well as involved in the precipitation of small amounts of DNA. It plays an important role as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, bases, oxidizing agents, reducing agents, iron and iron salts, copper, aluminum, brass and free radical initiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Melting Point 82°C to 85°C
Density 1.322
Boiling Point 125°C (25 mmHg)
Flash Point 138°C (280°F)
Odor Odorless
Quantity 500 g
UN Number UN2074
Beilstein 605349
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,129
Solubility Information Slightly soluble in water.
Formula Weight 71.08
Percent Purity ≥98%
Chemical Name or Material Acrylamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acrylamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H372-Causes damage to organs.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Acrylamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.